Product Details
- SNP ID
-
rs73330046
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:123501642 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGATATGTGGGTTGTGTGCTATTG[A/C]GGGCAGGGGCTGAAGCGGTGTTGTA
- Phenotype
-
MIM: 606604
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO32
PubMed Links
Gene Details
- Gene
- FBXO32
- Gene Name
- F-box protein 32
View Full Product Details