Product Details

SNP ID
rs73262317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66871519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCGTTCCTAATGCCCTGGATCA[A/G]GCCATCCAACTTGTAGGAATATCCC
Phenotype
MIM: 607591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C8orf44-SGK3 PubMed Links

Gene Details

Gene
C8orf44-SGK3
Gene Name
C8orf44-SGK3 readthrough
There are no transcripts associated with this gene.

Gene
MCMDC2
Gene Name
minichromosome maintenance domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136160.1 18 UTR 5 NP_001129632.1
NM_001136161.1 18 Intron NP_001129633.1
NM_173518.4 18 Intron NP_775789.3
XM_005251174.2 18 Intron XP_005251231.1
XM_006716427.3 18 Intron XP_006716490.1
XM_006716429.2 18 Intron XP_006716492.1
XM_006716433.3 18 Intron XP_006716496.1
XM_011517467.2 18 Intron XP_011515769.1
XM_011517468.2 18 Intron XP_011515770.1
XM_011517469.2 18 Intron XP_011515771.1
XM_017013140.1 18 Intron XP_016868629.1
Gene
SGK3
Gene Name
serum/glucocorticoid regulated kinase family member 3
There are no transcripts associated with this gene.

View Full Product Details