Product Details

SNP ID
rs73520817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:106868053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGAGACTATTTCCATGACTG[A/C]AAGCTAAAATCCAGTGAACTGAAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105376204 PubMed Links

Gene Details

Gene
LOC105376204
Gene Name
uncharacterized LOC105376204
There are no transcripts associated with this gene.

Gene
ZNF462
Gene Name
zinc finger protein 462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021224.4 Intron NP_067047.4
XM_006717209.3 Intron XP_006717272.1
XM_006717211.3 Intron XP_006717274.1
XM_006717212.3 Intron XP_006717275.1
XM_006717215.3 Intron XP_006717278.1
XM_006717216.3 Intron XP_006717279.1
XM_006717218.3 Intron XP_006717281.1
XM_011518892.2 Intron XP_011517194.1
XM_017014996.1 Intron XP_016870485.1
XM_017014997.1 Intron XP_016870486.1
XM_017014998.1 Intron XP_016870487.1
XM_017014999.1 Intron XP_016870488.1

View Full Product Details