Product Details

SNP ID
rs73656280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:111636756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTACCTGGAACAAAACTCTTAC[C/T]TTTTAAGAGTTTTGAGACCTTTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC25 PubMed Links

Gene Details

Gene
DNAJC25
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015882.2 Intron NP_001015882.2
Gene
DNAJC25-GNG10
Gene Name
DNAJC25-GNG10 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004125.3 Intron NP_004116.2

View Full Product Details