Product Details

SNP ID
rs73538608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96817391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGTAGGTTAAAATGAGGGTTG[A/T]CATGGCAGAAGAATTTCCCACATCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF782 PubMed Links

Gene Details

Gene
ZNF782
Gene Name
zinc finger protein 782
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001662.1 3111 UTR 3 NP_001001662.1
XM_005251742.4 3111 UTR 3 XP_005251799.1
XM_005251744.3 3111 UTR 3 XP_005251801.1
XM_011518315.2 3111 UTR 3 XP_011516617.1
XM_011518318.2 3111 UTR 3 XP_011516620.1
XM_011518320.2 3111 UTR 3 XP_011516622.1
XM_017014343.1 3111 UTR 3 XP_016869832.1
XM_017014344.1 3111 UTR 3 XP_016869833.1

View Full Product Details