Product Details
- SNP ID
-
rs71452030
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:26911235 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTCAACAAGGGGTTTTTTGTTTG[A/G]AGGTTCTGGGGAATCAGGCGAATCT
- Phenotype
-
MIM: 615079
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ASUN
PubMed Links
Gene Details
- Gene
- ASUN
- Gene Name
- asunder, spermatogenesis regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018164.2 |
1958 |
Missense Mutation |
CCA,TCA |
P630S |
NP_060634.2 |
XM_005253422.3 |
1958 |
Missense Mutation |
CCA,TCA |
P573S |
XP_005253479.1 |
XM_011520750.2 |
1958 |
Missense Mutation |
CCA,TCA |
P636S |
XP_011519052.1 |
XM_011520751.2 |
1958 |
Missense Mutation |
CCA,TCA |
P579S |
XP_011519053.1 |
XM_011520752.2 |
1958 |
Missense Mutation |
CCA,TCA |
P561S |
XP_011519054.1 |
XM_011520753.2 |
1958 |
Missense Mutation |
CCA,TCA |
P358S |
XP_011519055.1 |
XM_017019632.1 |
1958 |
Missense Mutation |
CCA,TCA |
P630S |
XP_016875121.1 |
XM_017019633.1 |
1958 |
Missense Mutation |
CCA,TCA |
P611S |
XP_016875122.1 |
XM_017019634.1 |
1958 |
Missense Mutation |
CCA,TCA |
P605S |
XP_016875123.1 |
XM_017019635.1 |
1958 |
Missense Mutation |
CCA,TCA |
P573S |
XP_016875124.1 |
XM_017019636.1 |
1958 |
Missense Mutation |
CCA,TCA |
P555S |
XP_016875125.1 |
XM_017019637.1 |
1958 |
Missense Mutation |
CCA,TCA |
P352S |
XP_016875126.1 |
View Full Product Details