Product Details
- SNP ID
-
rs71541561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120213914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAGTTCTCCAGGATGGCCCGGGC[A/G]CAGCCGCCACACTTGGGTGCGAACA
- Phenotype
-
MIM: 602505
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PXN
PubMed Links
Gene Details
- Gene
- PXN
- Gene Name
- paxillin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080855.2 |
2260 |
Silent Mutation |
TGC,TGT |
C479C |
NP_001074324.1 |
NM_001243756.1 |
2260 |
Silent Mutation |
TGC,TGT |
C493C |
NP_001230685.1 |
NM_002859.3 |
2260 |
Silent Mutation |
TGC,TGT |
C445C |
NP_002850.2 |
NM_025157.4 |
2260 |
Silent Mutation |
TGC,TGT |
C312C |
NP_079433.3 |
XM_005253917.3 |
2260 |
Silent Mutation |
TGC,TGT |
C291C |
XP_005253974.1 |
XM_006719532.1 |
2260 |
Silent Mutation |
TGC,TGT |
C1017C |
XP_006719595.1 |
XM_006719534.1 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_006719597.1 |
XM_006719535.3 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_006719598.1 |
XM_006719536.2 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_006719599.1 |
XM_011538622.1 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_011536924.1 |
XM_011538623.1 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_011536925.1 |
XM_017019726.1 |
2260 |
Silent Mutation |
TGC,TGT |
C1023C |
XP_016875215.1 |
XM_017019727.1 |
2260 |
Silent Mutation |
TGC,TGT |
C1018C |
XP_016875216.1 |
XM_017019728.1 |
2260 |
Silent Mutation |
TGC,TGT |
C1015C |
XP_016875217.1 |
XM_017019729.1 |
2260 |
Silent Mutation |
TGC,TGT |
C975C |
XP_016875218.1 |
XM_017019730.1 |
2260 |
Silent Mutation |
TGC,TGT |
C1023C |
XP_016875219.1 |
XM_017019731.1 |
2260 |
Silent Mutation |
TGC,TGT |
C884C |
XP_016875220.1 |
XM_017019732.1 |
2260 |
Intron |
|
|
XP_016875221.1 |
XM_017019733.1 |
2260 |
Silent Mutation |
TGC,TGT |
C828C |
XP_016875222.1 |
XM_017019734.1 |
2260 |
Silent Mutation |
TGC,TGT |
C776C |
XP_016875223.1 |
XM_017019735.1 |
2260 |
Silent Mutation |
TGC,TGT |
C746C |
XP_016875224.1 |
XM_017019736.1 |
2260 |
Silent Mutation |
TGC,TGT |
C636C |
XP_016875225.1 |
XM_017019737.1 |
2260 |
Silent Mutation |
TGC,TGT |
C588C |
XP_016875226.1 |
XM_017019738.1 |
2260 |
Silent Mutation |
TGC,TGT |
C533C |
XP_016875227.1 |
XM_017019739.1 |
2260 |
Silent Mutation |
TGC,TGT |
C499C |
XP_016875228.1 |
XM_017019740.1 |
2260 |
Silent Mutation |
TGC,TGT |
C485C |
XP_016875229.1 |
XM_017019741.1 |
2260 |
Silent Mutation |
TGC,TGT |
C477C |
XP_016875230.1 |
XM_017019742.1 |
2260 |
Silent Mutation |
TGC,TGT |
C451C |
XP_016875231.1 |
XM_017019743.1 |
2260 |
Silent Mutation |
TGC,TGT |
C312C |
XP_016875232.1 |
- Gene
- PXN-AS1
- Gene Name
- PXN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details