Product Details

SNP ID
rs202225683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109117047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCTGTTTCCTCAAATGGTTGTT[C/T]TTTTTTTTTTTTTTTTTTTTTTTAA
Phenotype
MIM: 601557 MIM: 191525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACACB PubMed Links

Gene Details

Gene
ACACB
Gene Name
acetyl-CoA carboxylase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093.3 Intron NP_001084.3
XM_005253876.4 Intron XP_005253933.1
XM_006719367.3 Intron XP_006719430.1
XM_011538259.2 Intron XP_011536561.1
XM_011538263.2 Intron XP_011536565.1
XM_011538264.2 Intron XP_011536566.1
XM_011538265.2 Intron XP_011536567.1
XM_017019252.1 Intron XP_016874741.1
Gene
LOC105369974
Gene Name
uncharacterized LOC105369974
There are no transcripts associated with this gene.

Gene
UNG
Gene Name
uracil DNA glycosylase
There are no transcripts associated with this gene.

View Full Product Details