Product Details

SNP ID
rs73191829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110511101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGATGGAGGGTACTGAAAGAG[A/G]GTAATAGAGATATGATCTGTAGTCT
Phenotype
MIM: 608368 MIM: 606932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD9B PubMed Links

Gene Details

Gene
RAD9B
Gene Name
RAD9 checkpoint clamp component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286531.1 Intron NP_001273460.1
NM_001286532.1 Intron NP_001273461.1
NM_001286533.1 Intron NP_001273462.1
NM_001286534.1 Intron NP_001273463.1
NM_001286535.1 Intron NP_001273464.1
NM_001286536.1 Intron NP_001273465.1
NM_152442.3 Intron NP_689655.3
XM_005253849.4 Intron XP_005253906.1
XM_011537971.2 Intron XP_011536273.1
XM_011537972.2 Intron XP_011536274.1
XM_011537974.2 Intron XP_011536276.1
XM_017018877.1 Intron XP_016874366.1
XM_017018878.1 Intron XP_016874367.1
XM_017018879.1 Intron XP_016874368.1
XM_017018880.1 Intron XP_016874369.1
XM_017018881.1 Intron XP_016874370.1
XM_017018882.1 Intron XP_016874371.1
Gene
VPS29
Gene Name
VPS29, retromer complex component
There are no transcripts associated with this gene.

View Full Product Details