Product Details

SNP ID
rs73425095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112794317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGTTCTCTGACTATGAGATAG[A/G]TATGTGTGTGAGTGTGAGTGACTGC
Phenotype
MIM: 612159
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPH3A PubMed Links

Gene Details

Gene
RPH3A
Gene Name
rabphilin 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143854.1 Intron NP_001137326.1
NM_014954.3 Intron NP_055769.2
XM_011538048.1 Intron XP_011536350.1
XM_017019034.1 Intron XP_016874523.1

View Full Product Details