Product Details

SNP ID
rs72740860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58415682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATTAAATAATCTGACACTCCT[C/T]AAGTGTTGGTGATGAGGTGGAAAAA
Phenotype
MIM: 151670
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIPC PubMed Links

Gene Details

Gene
LIPC
Gene Name
lipase C, hepatic type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000236.2 Intron NP_000227.2
XM_005254372.1 Intron XP_005254429.1
XM_005254374.4 Intron XP_005254431.2
XM_006720502.3 Intron XP_006720565.1
XM_017022176.1 Intron XP_016877665.1
Gene
LOC101928635
Gene Name
uncharacterized LOC101928635
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011522293.2 Intron XP_011520595.1

View Full Product Details