Product Details

SNP ID
rs73438476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72285742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCATAACGTTCTCATTACCTGCA[C/T]GTGAGCCTAGACCTCAAATCCTTAA
Phenotype
MIM: 612681
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELF6 PubMed Links

Gene Details

Gene
CELF6
Gene Name
CUGBP, Elav-like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172684.1 2066 UTR 3 NP_001166155.1
NM_001172685.1 2066 UTR 3 NP_001166156.1
NM_052840.4 2066 UTR 3 NP_443072.3

View Full Product Details