Product Details

SNP ID
rs71383203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11269146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTCCGGCTGCCACGATGGCT[C/G]TGTCTGCAACTCTGGCTGAAGGTTT
Phenotype
MIM: 182890 MIM: 190232
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRM2 PubMed Links

Gene Details

Gene
PRM2
Gene Name
protamine 2
There are no transcripts associated with this gene.

Gene
PRM3
Gene Name
protamine 3
There are no transcripts associated with this gene.

Gene
TNP2
Gene Name
transition protein 2 (during histone to protamine replacement)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005425.4 158 Silent Mutation CAC,CAG H39Q NP_005416.1

View Full Product Details