Product Details

SNP ID
rs72788065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57472324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACAAAAAACCCCACACAACCAAA[A/G]GTTTGACGTGAATAGAAAGATAGCC
Phenotype
MIM: 608333 MIM: 180663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOK4 PubMed Links

Gene Details

Gene
DOK4
Gene Name
docking protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018110.3 2443 UTR 3 NP_060580.2
XM_005256042.3 2443 UTR 3 XP_005256099.1
XM_005256043.2 2443 UTR 3 XP_005256100.1
Gene
POLR2C
Gene Name
RNA polymerase II subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032940.2 2443 Intron NP_116558.1

View Full Product Details