Product Details

SNP ID
rs72825269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81101069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTATAGAGTCGCTGGTAAGCGTA[C/T]GCCCGAGGCCCTGGGCGTCCCCACT
Phenotype
MIM: 238330 MIM: 607894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCSH PubMed Links

Gene Details

Gene
GCSH
Gene Name
glycine cleavage system protein H
There are no transcripts associated with this gene.

Gene
PKD1L2
Gene Name
polycystin 1 like 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076780.1 7434 Intron NP_001070248.1
NM_001278423.1 7434 Intron NP_001265352.1
NM_001278425.1 7434 UTR 3 NP_001265354.1
NM_052892.3 7434 UTR 3 NP_443124.3

View Full Product Details