Product Details

SNP ID
rs72811442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88680941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTGGCCTCTAGTCTGTGGTCT[A/G]TGGGACTTTGTACAGATAAGTCAAA
Phenotype
MIM: 612741
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNAI3 PubMed Links

Gene Details

Gene
SNAI3
Gene Name
snail family transcriptional repressor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178310.3 Intron NP_840101.1
Gene
SNAI3-AS1
Gene Name
SNAI3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details