Product Details
- SNP ID
-
rs17843023
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40960351 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CATTCCAGCCGGGACTTGACGTCCA[G/T]CAGGATCTCGTATTCTTGGTTCTGT
- Phenotype
-
MIM: 616678
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT39
PubMed Links
Gene Details
- Gene
- KRT39
- Gene Name
- keratin 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213656.3 |
1183 |
Missense Mutation |
ATG,CTG |
M383L |
NP_998821.3 |
View Full Product Details