Product Details

SNP ID
rs72817881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34586771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTAGATAATTCAATGCTTGTTG[A/C]ACGAATGAATGAGTGCAAGGATGAA
Phenotype
MIM: 616178
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C17orf102 PubMed Links

Gene Details

Gene
C17orf102
Gene Name
chromosome 17 open reading frame 102
There are no transcripts associated with this gene.

Gene
TMEM132E
Gene Name
transmembrane protein 132E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304438.1 Intron NP_001291367.1

View Full Product Details