Product Details

SNP ID
rs73423664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:32514511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTTCAAAAAAATAAAAAAGTTT[A/C]TTTTTGTCTAGAGTCAAATGAATGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WBP11P1 PubMed Links

Gene Details

Gene
WBP11P1
Gene Name
WW domain binding protein 11 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details