Product Details

SNP ID
rs73027454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37885463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAGGAAACCTGCGTTTATCCTT[A/G]AAATCCCAGAAGTCCTTTTGAAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR87 PubMed Links

Gene Details

Gene
WDR87
Gene Name
WD repeat domain 87
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291088.1 8462 Silent Mutation TTC,TTT F2736F NP_001278017.1
NM_031951.4 8462 Silent Mutation TTC,TTT F2697F NP_114157.4
XM_011527360.2 8462 Silent Mutation TTC,TTT F2736F XP_011525662.1

View Full Product Details