Product Details

SNP ID
rs73607034
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55352072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGGTGTGGCACCCTGTGCTTG[A/G]GATGTTACCTGGGTGACCTCACCAA
Phenotype
MIM: 613198
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
COX6B2 PubMed Links

Gene Details

Gene
COX6B2
Gene Name
cytochrome c oxidase subunit 6B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144613.4 Intron NP_653214.2
Gene
FAM71E2
Gene Name
family with sequence similarity 71 member E2
There are no transcripts associated with this gene.

Gene
KMT5C
Gene Name
lysine methyltransferase 5C
There are no transcripts associated with this gene.

View Full Product Details