Product Details

SNP ID
rs71354061
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57855891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGTGGACCTTTTTATGTCAGGA[A/C]CTCAGCATGGATACCTATTTGTCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF587 PubMed Links

Gene Details

Gene
ZNF587
Gene Name
zinc finger protein 587
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204817.1 Intron NP_001191746.1
NM_032828.3 Intron NP_116217.1
Gene
ZNF587B
Gene Name
zinc finger protein 587B
There are no transcripts associated with this gene.

View Full Product Details