Product Details

SNP ID
rs112608565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21408488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCAGAATATTTTATTTATAATTA[C/T]ACTGCATATTCTCTGAAATTTTACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF493 PubMed Links

Gene Details

Gene
ZNF493
Gene Name
zinc finger protein 493
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076678.2 1391 Intron NP_001070146.1
NM_145326.2 1391 UTR 3 NP_663299.2
NM_175910.6 1391 Intron NP_787106.4

View Full Product Details