Product Details
- SNP ID
-
rs73121283
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:41046877 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAGAAGCAGAATTAGCAGTGGGC[C/T]AGTCATTGGCCTCTCTTCAGAGTTC
- Phenotype
-
MIM: 126420
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TOP1
PubMed Links
Gene Details
- Gene
- TOP1
- Gene Name
- topoisomerase (DNA) I
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003286.2 |
|
Intron |
|
|
NP_003277.1 |
View Full Product Details