Product Details

SNP ID
rs73569630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155062909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCATGGATGAAAGGCTTAATAC[A/G]TATAAAGTCCTTATAACGGTGCTTG
Phenotype
MIM: 300617 MIM: 300116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRCC3 PubMed Links

Gene Details

Gene
BRCC3
Gene Name
BRCA1/BRCA2-containing complex subunit 3
There are no transcripts associated with this gene.

Gene
CMC4
Gene Name
C-X9-C motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018024.2 Intron NP_001018024.1
Gene
FUNDC2
Gene Name
FUN14 domain containing 2
There are no transcripts associated with this gene.

Gene
MTCP1
Gene Name
mature T-cell proliferation 1
There are no transcripts associated with this gene.

View Full Product Details