Product Details

SNP ID
rs72625303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12913012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGATTAACTTTGAGAAATATGAT[A/G]ATATACCAGTAGAGGCAACCGGCAG
Phenotype
MIM: 400010
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX3Y PubMed Links

Gene Details

Gene
DDX3Y
Gene Name
DEAD-box helicase 3, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122665.3 905 Missense Mutation AAT,GAT N163D NP_001116137.1
NM_001302552.2 905 Intron NP_001289481.1
NM_001324195.1 905 Intron NP_001311124.1
NM_004660.4 905 Intron NP_004651.2

View Full Product Details