Product Details

SNP ID
rs73837570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86942254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAAATGAGGACAATGCCTTGTA[C/T]GATATTTTACCAGCCCACAGGGAAT
Phenotype
MIM: 159557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AFF1 PubMed Links

Gene Details

Gene
AFF1
Gene Name
AF4/FMR2 family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166693.2 Intron NP_001160165.1
NM_001313959.1 Intron NP_001300888.1
NM_001313960.1 Intron NP_001300889.1
NM_005935.3 Intron NP_005926.1
XM_005263007.3 Intron XP_005263064.1
XM_005263009.3 Intron XP_005263066.1
XM_005263011.3 Intron XP_005263068.1
XM_005263013.3 Intron XP_005263070.1
XM_011531973.2 Intron XP_011530275.1
XM_017008215.1 Intron XP_016863704.1
XM_017008216.1 Intron XP_016863705.1
Gene
LOC100506746
Gene Name
uncharacterized LOC100506746
There are no transcripts associated with this gene.

View Full Product Details