Product Details

SNP ID
rs72684898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:101791500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTTACTAACTTGGTCAAAGGA[A/G]AACGGTCAACTTGGTTGAATGCAGA
Phenotype
MIM: 610292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BANK1 PubMed Links

Gene Details

Gene
BANK1
Gene Name
B-cell scaffold protein with ankyrin repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083907.2 Intron NP_001077376.2
NM_001127507.2 Intron NP_001120979.2
NM_017935.4 Intron NP_060405.4
XM_017008337.1 Intron XP_016863826.1

View Full Product Details