Product Details

SNP ID
rs71463857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80137251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATAATTTCATGTTATATAATA[A/G]TATCTCGGTAGCAAAGAAAAAAACC
Phenotype
MIM: 614925
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTOGL PubMed Links

Gene Details

Gene
OTOGL
Gene Name
otogelin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173591.3 Intron NP_775862.3
XM_005268802.3 Intron XP_005268859.1
XM_011538191.1 Intron XP_011536493.1
XM_011538192.2 Intron XP_011536494.1
XM_011538193.1 Intron XP_011536495.1

View Full Product Details