Product Details

SNP ID
rs73097943
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:219914120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTAAGAGATGGTTATTTTCAG[A/G]CAATCTTGCAGTCTACCAGAGATGA
Phenotype
MIM: 611146
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC30A10 PubMed Links

Gene Details

Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018713.2 3334 Intron NP_061183.2
XM_006711437.3 3334 UTR 3 XP_006711500.2
XM_017001684.1 3334 UTR 3 XP_016857173.1

View Full Product Details