Product Details

SNP ID
rs74140013
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:212438128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAGATAACAGGTTATAAAGTGA[A/G]TGTGTTGAACAAAATTGAAAATAGA
Phenotype
MIM: 611874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NENF PubMed Links

Gene Details

Gene
NENF
Gene Name
neudesin neurotrophic factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013349.4 Intron NP_037481.1

View Full Product Details