Product Details

SNP ID
rs72632594
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:6270817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCTGAGGCCAAGTGCAGCCTGG[A/G]AACTGCTCCAAGGACTGTCCTGCTG
Phenotype
MIM: 602587 MIM: 614269
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT7 PubMed Links

Gene Details

Gene
ACOT7
Gene Name
acyl-CoA thioesterase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007274.3 Intron NP_009205.3
NM_181864.2 Intron NP_863654.1
NM_181865.2 Intron NP_863655.1
NM_181866.2 Intron NP_863656.1
Gene
GPR153
Gene Name
G protein-coupled receptor 153
There are no transcripts associated with this gene.

View Full Product Details