Product Details

SNP ID
rs73952503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:108542975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGCATATGTGAAAGTGATATT[G/T]ACAAATTTTGGAACACATCTTTCAA
Phenotype
MIM: 602567
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GCC2-AS1 PubMed Links

Gene Details

Gene
GCC2-AS1
Gene Name
GCC2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
LIMS1
Gene Name
LIM zinc finger domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193482.1 Intron NP_001180411.1
NM_001193483.2 Intron NP_001180412.1
NM_001193484.1 Intron NP_001180413.1
NM_001193485.2 Intron NP_001180414.1
NM_001193488.1 Intron NP_001180417.1
NM_004987.5 Intron NP_004978.2
XM_005263946.1 Intron XP_005264003.1
XM_005263948.3 Intron XP_005264005.1
XM_005263949.1 Intron XP_005264006.1
XM_005263950.1 Intron XP_005264007.1
XM_005263951.4 Intron XP_005264008.1
XM_017004091.1 Intron XP_016859580.1
XM_017004092.1 Intron XP_016859581.1
XM_017004093.1 Intron XP_016859582.1
XM_017004094.1 Intron XP_016859583.1
XM_017004095.1 Intron XP_016859584.1
XM_017004096.1 Intron XP_016859585.1

View Full Product Details