Product Details

SNP ID
rs72910670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69960120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAATCAATCTTTATTGGTGTTT[A/G]CAGCAATTGAATTATTGGCATAAGG
Phenotype
MIM: 611765
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASPRV1 PubMed Links

Gene Details

Gene
ASPRV1
Gene Name
aspartic peptidase, retroviral-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152792.2 2146 UTR 3 NP_690005.2
Gene
PCBP1-AS1
Gene Name
PCBP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details