Product Details

SNP ID
rs72908583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74838853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCTGGCTGATTCTTTCTATTGA[C/T]TGAGATAAGTTAGTATACAAGATGC
Phenotype
MIM: 601125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HK2 PubMed Links

Gene Details

Gene
HK2
Gene Name
hexokinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000189.4 Intron NP_000180.2
XM_005264280.1 Intron XP_005264337.1
XM_011532807.1 Intron XP_011531109.1
XM_017003945.1 Intron XP_016859434.1

View Full Product Details