Product Details

SNP ID
rs72831579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85422634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTCAAGGGTTTTAGGCAGAGCC[A/G]TGACACAACCAGAGATACTACGAGC
Phenotype
MIM: 153615
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPG PubMed Links

Gene Details

Gene
CAPG
Gene Name
capping actin protein, gelsolin like
There are no transcripts associated with this gene.

Gene
SH2D6
Gene Name
SH2 domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201594.2 519 UTR 5 NP_963888.2
XM_011532783.1 519 Intron XP_011531085.1
XM_011532796.2 519 Intron XP_011531098.1
XM_017003834.1 519 Intron XP_016859323.1
XM_017003835.1 519 Intron XP_016859324.1
XM_017003836.1 519 Intron XP_016859325.1
XM_017003837.1 519 Intron XP_016859326.1
XM_017003838.1 519 Intron XP_016859327.1
XM_017003839.1 519 Intron XP_016859328.1
XM_017003840.1 519 Intron XP_016859329.1
XM_017003841.1 519 Intron XP_016859330.1
XM_017003842.1 519 Intron XP_016859331.1
XM_017003843.1 519 Intron XP_016859332.1
XM_017003844.1 519 Intron XP_016859333.1
XM_017003845.1 519 Intron XP_016859334.1
XM_017003846.1 519 Intron XP_016859335.1
XM_017003847.1 519 Intron XP_016859336.1
XM_017003848.1 519 Intron XP_016859337.1
XM_017003849.1 519 Intron XP_016859338.1
XM_017003850.1 519 Intron XP_016859339.1
XM_017003851.1 519 Intron XP_016859340.1
XM_017003852.1 519 Intron XP_016859341.1
XM_017003853.1 519 Intron XP_016859342.1
XM_017003854.1 519 Intron XP_016859343.1
XM_017003855.1 519 Intron XP_016859344.1
XM_017003856.1 519 Intron XP_016859345.1
XM_017003857.1 519 Intron XP_016859346.1
XM_017003858.1 519 UTR 5 XP_016859347.1
XM_017003859.1 519 Intron XP_016859348.1
XM_017003860.1 519 Intron XP_016859349.1

View Full Product Details