Product Details

SNP ID
rs73038350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:18348230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATCTATTAGTTTTATTTACCT[A/G]AGCTTATTTGCATGATAGTAAAAAT
Phenotype
MIM: 602075
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SATB1 PubMed Links

Gene Details

Gene
SATB1
Gene Name
SATB homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001131010.3 3513 UTR 3 NP_001124482.1
NM_001195470.2 3513 UTR 3 NP_001182399.1
NM_001322871.1 3513 UTR 3 NP_001309800.1
NM_001322872.1 3513 UTR 3 NP_001309801.1
NM_001322873.1 3513 UTR 3 NP_001309802.1
NM_001322874.1 3513 UTR 3 NP_001309803.1
NM_001322875.1 3513 UTR 3 NP_001309804.1
NM_001322876.1 3513 UTR 3 NP_001309805.1
NM_002971.5 3513 UTR 3 NP_002962.1
XM_011533988.2 3513 Intron XP_011532290.1
XM_011533989.2 3513 Intron XP_011532291.1
XM_011533990.2 3513 Intron XP_011532292.1

View Full Product Details