Product Details

SNP ID
rs73820942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:21425480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTGGTTTTCATTCCTAGAATA[A/C]GTGCTGTTTACCTGTTTAAGTTGCG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF385D PubMed Links

Gene Details

Gene
ZNF385D
Gene Name
zinc finger protein 385D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024697.2 Intron NP_078973.1
XM_011534122.1 Intron XP_011532424.1
XM_011534123.2 Intron XP_011532425.1
XM_011534124.2 Intron XP_011532426.1
XM_017007191.1 Intron XP_016862680.1
XM_017007192.1 Intron XP_016862681.1
XM_017007193.1 Intron XP_016862682.1
XM_017007194.1 Intron XP_016862683.1
XM_017007195.1 Intron XP_016862684.1
XM_017007196.1 Intron XP_016862685.1
XM_017007197.1 Intron XP_016862686.1
XM_017007198.1 Intron XP_016862687.1
XM_017007199.1 Intron XP_016862688.1
XM_017007200.1 Intron XP_016862689.1
XM_017007201.1 Intron XP_016862690.1
XM_017007202.1 Intron XP_016862691.1
XM_017007203.1 Intron XP_016862692.1

View Full Product Details