Product Details

SNP ID
rs73087706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58493627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTCTGGAGCTCTGGGAAAAGG[A/G]GGACCTGGGGAGATTTGAAGCAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD6 PubMed Links

Gene Details

Gene
KCTD6
Gene Name
potassium channel tetramerization domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128214.1 625 Intron NP_001121686.1
NM_153331.3 625 Intron NP_699162.3
XM_005264937.2 625 UTR 5 XP_005264994.1

View Full Product Details