Product Details

SNP ID
rs71503679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69826034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATAGGAAGAAACCAGAAAGATAA[A/G]CAGAAAAAAAGGGAAGAATTATGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf135 PubMed Links

Gene Details

Gene
C9orf135
Gene Name
chromosome 9 open reading frame 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010940.2 Intron NP_001010940.1
NM_001308084.1 Intron NP_001295013.1
NM_001308085.1 Intron NP_001295014.1
NM_001308086.1 Intron NP_001295015.1
XM_011518229.2 Intron XP_011516531.1
XM_011518230.2 Intron XP_011516532.1
XM_011518231.2 Intron XP_011516533.1
XM_011518233.2 Intron XP_011516535.1
XM_011518234.2 Intron XP_011516536.1
Gene
C9orf135-AS1
Gene Name
C9orf135 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details