Product Details

SNP ID
rs72956167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:59224731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTTTGGCCTCCTTAACAGCAG[C/T]TTTTTAAAAAAATTATTTACAAGGC
Phenotype
MIM: 137260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRP PubMed Links

Gene Details

Gene
GRP
Gene Name
gastrin releasing peptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012512.2 Intron NP_001012530.1
NM_001012513.2 Intron NP_001012531.1
NM_002091.4 Intron NP_002082.2
XM_011525933.2 Intron XP_011524235.1
XM_017025712.1 Intron XP_016881201.1
XM_017025713.1 Intron XP_016881202.1
XM_017025714.1 Intron XP_016881203.1

View Full Product Details