Product Details
- SNP ID
-
rs72546304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:143941682 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGCCCAACAGGGGGGACAGGTC[C/T]AGAGAGGCAGGTATTGTTAGGTTCA
- Phenotype
-
MIM: 603044
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLAGL1
PubMed Links
Gene Details
- Gene
- PLAGL1
- Gene Name
- PLAG1 like zinc finger 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001080951.2 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001074420.1 |
NM_001080952.2 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001074421.1 |
NM_001080953.2 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001074422.1 |
NM_001080954.2 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001074423.1 |
NM_001080955.2 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001074424.1 |
NM_001080956.2 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001074425.1 |
NM_001289037.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001275966.1 |
NM_001289038.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001275967.1 |
NM_001289039.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001275968.1 |
NM_001289040.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001275969.1 |
NM_001289041.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001275970.1 |
NM_001289042.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275971.1 |
NM_001289043.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275972.1 |
NM_001289044.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275973.1 |
NM_001289045.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275974.1 |
NM_001289046.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275975.1 |
NM_001289047.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275976.1 |
NM_001289048.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275977.1 |
NM_001289049.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001275978.1 |
NM_001317156.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001304085.1 |
NM_001317157.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001304086.1 |
NM_001317158.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001304087.1 |
NM_001317159.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001304088.1 |
NM_001317160.1 |
1817 |
Silent Mutation |
CTA,CTG |
L326L |
NP_001304089.1 |
NM_001317161.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001304090.1 |
NM_001317162.1 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_001304091.1 |
NM_006718.4 |
1817 |
Silent Mutation |
CTA,CTG |
L378L |
NP_006709.2 |
- Gene
- ZC2HC1B
- Gene Name
- zinc finger C2HC-type containing 1B
There are no transcripts associated with this gene.
View Full Product Details