Product Details

SNP ID
rs73292095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:53040349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAATGTGGCAGCCACAGCTGATG[A/C]ATAGGGAGATCTTTCAATTTTCATA
Phenotype
MIM: 614603
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DDHD1 PubMed Links

Gene Details

Gene
DDHD1
Gene Name
DDHD domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160147.1 9352 UTR 3 NP_001153619.1
NM_001160148.1 9352 UTR 3 NP_001153620.1
NM_030637.2 9352 UTR 3 NP_085140.2
XM_005268102.2 9352 Intron XP_005268159.1
XM_005268103.2 9352 Intron XP_005268160.1
XM_005268105.2 9352 Intron XP_005268162.1
XM_011537188.2 9352 UTR 3 XP_011535490.1
XM_011537189.2 9352 Intron XP_011535491.1
XM_017021668.1 9352 Intron XP_016877157.1
XM_017021669.1 9352 Intron XP_016877158.1

View Full Product Details