Product Details

SNP ID
rs74065407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70725424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCACCTGGCCTGCACGGCTAACC[A/G]TAAGTCCCGGGCTGAGTCATGATAA
Phenotype
MIM: 600136
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K9 PubMed Links

Gene Details

Gene
MAP3K9
Gene Name
mitogen-activated protein kinase kinase kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284230.1 8297 UTR 3 NP_001271159.1
NM_001284231.1 8297 UTR 3 NP_001271160.1
NM_001284232.1 8297 UTR 3 NP_001271161.1
NM_033141.3 8297 UTR 3 NP_149132.2
XM_005267683.4 8297 UTR 3 XP_005267740.1
XM_011536788.2 8297 UTR 3 XP_011535090.1
XM_011536792.1 8297 UTR 3 XP_011535094.1
XM_011536794.2 8297 UTR 3 XP_011535096.1

View Full Product Details