Product Details

SNP ID
rs72631832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100307866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAATGTTAAGCAGAGACCCAAA[C/T]CCTAGGTCTGCTGACTCCTAGTCCA
Phenotype
MIM: 615064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR345 PubMed Links

Gene Details

Gene
MIR345
Gene Name
microRNA 345
There are no transcripts associated with this gene.

Gene
SLC25A29
Gene Name
solute carrier family 25 member 29
There are no transcripts associated with this gene.

View Full Product Details