Product Details
- SNP ID
-
rs74096642
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52821995 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGCTGAATCCACCCTTGGTGGC[C/T]CCCCCACTCCCACCCAGGGAGATGC
- Phenotype
-
MIM: 123940
MIM: 611160
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT4
PubMed Links
Gene Details
- Gene
- KRT4
- Gene Name
- keratin 4
There are no transcripts associated with this gene.
- Gene
- KRT79
- Gene Name
- keratin 79
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175834.2 |
1518 |
Silent Mutation |
GGA,GGG |
G495G |
NP_787028.1 |
View Full Product Details