Product Details

SNP ID
rs71459343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56106623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTGGTGGAAGCATCTAGCTCA[A/G]GTGTGTCGGTACTGAGCCTGTGTGA
Phenotype
MIM: 190151 MIM: 602145 MIM: 613315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERBB3 PubMed Links

Gene Details

Gene
ERBB3
Gene Name
erb-b2 receptor tyrosine kinase 3
There are no transcripts associated with this gene.

Gene
PA2G4
Gene Name
proliferation-associated 2G4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006191.2 543 Missense Mutation AGT,GGT S42G NP_006182.2
Gene
RPL41
Gene Name
ribosomal protein L41
There are no transcripts associated with this gene.

View Full Product Details