Product Details

SNP ID
rs73324304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56239190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCCCATCTCTGGGATTTGGTC[A/G]CTTCTCTAGGGGTTGGGTTGGGAGG
Phenotype
MIM: 612104 MIM: 608730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173595.3 7273 UTR 3 NP_775866.2
XM_011538197.2 7273 Intron XP_011536499.1
XM_017019183.1 7273 Intron XP_016874672.1
XM_017019184.1 7273 Intron XP_016874673.1
Gene
NABP2
Gene Name
nucleic acid binding protein 2
There are no transcripts associated with this gene.

Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
There are no transcripts associated with this gene.

View Full Product Details