Product Details

SNP ID
rs72559704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104254948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACGATGTCCGGGGAGTCAGCCA[A/G]GAGCTTGGGGAAGGGTGAGGCCTGC
Phenotype
MIM: 605482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTO1 PubMed Links

Gene Details

Gene
GSTO1
Gene Name
glutathione S-transferase omega 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191002.1 239 Missense Mutation AAG,AGG K7R NP_001177931.1
NM_001191003.1 239 Intron NP_001177932.1
NM_004832.2 239 Missense Mutation AAG,AGG K7R NP_004823.1

View Full Product Details