Product Details
- SNP ID
-
rs72550759
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59855951 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCAGGTAAGACCTGGGCTGCAGC[G/A]TTTGGATTGCTGAATGCTCCTTGAG
- Phenotype
-
MIM: 189905
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCN1
PubMed Links
Gene Details
- Gene
- TCN1
- Gene Name
- transcobalamin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001062.3 |
953 |
Silent Mutation |
AAC,AAT |
N285N |
NP_001053.2 |
View Full Product Details